SY3H3D67FC1B Synthonix Corporation

Methyl 2-(2-amino-5-methylthiazol-4-yl)acetate

  • MDL number No MDL found
  • Cas Number : 259654-73-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/43675.html

Safety Information

MSDS https://synthonix.com/43675.html
Availability details for
Quantity
  • Availability
  • Add to Cart