SY3H3D67FC1C Synthonix Corporation

Methyl 3-methoxy-1-methyl-1H-pyrazole-5-carboxylate

  • MDL number No MDL found
  • Cas Number : 55781-86-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/43679.html

Safety Information

MSDS https://synthonix.com/43679.html
Availability details for
Quantity
  • Availability
  • Add to Cart