SY3H3D67FC1D Synthonix Corporation

Methyl 5-methoxy-1-methyl-1H-pyrazole-3-carboxylate

  • MDL number No MDL found
  • Cas Number : 58364-91-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/43680.html

Safety Information

MSDS https://synthonix.com/43680.html
Availability details for
Quantity
  • Availability
  • Add to Cart