SY3H3D67FC1E Synthonix Corporation

3-(4-Methyl-2,5-dioxoimidazolidin-4-yl)propanoic acid

  • MDL number No MDL found
  • Cas Number : 43189-50-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/43683.html

Safety Information

MSDS https://synthonix.com/43683.html
Availability details for
Quantity
  • Availability
  • Add to Cart