SY3H3D67FC1F Synthonix Corporation

4-Methoxy-6-methyl-2-(methylthio)pyrimidine

  • Cas Number : 55749-33-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43685.html

Safety Information

MSDS https://synthonix.com/43685.html
Availability details for
Quantity
  • Availability
  • Add to Cart