SY3H3D67FC22 Synthonix Corporation

5-Methyl-4,5,6,7-tetrahydrothiazolo[5,4-c]pyridine

  • Cas Number : 259809-24-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43689.html

Safety Information

MSDS https://synthonix.com/43689.html
Availability details for
Quantity
  • Availability
  • Add to Cart