SY3H3D67FC28 Synthonix Corporation

Methyl 3,4-Dihydro-2H-pyran-5-carboxylate

  • Cas Number : 86971-83-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/43727.html

Safety Information

MSDS https://synthonix.com/43727.html
Availability details for
Quantity
  • Availability
  • Add to Cart