SY3H3D67FC2E Synthonix Corporation

Methyl 2-(3,5-dimethyl-1H-1,2,4-triazol-1-yl)acetate

  • MDL number No MDL found
  • Cas Number : 1250278-88-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/43761.html

Safety Information

MSDS https://synthonix.com/43761.html
Availability details for
Quantity
  • Availability
  • Add to Cart