SY3H3D67FC31 Synthonix Corporation

Methyl 2-oxopiperidine-3-carboxylate

  • MDL number No MDL found
  • Cas Number : 106118-94-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43779.html

Safety Information

MSDS https://synthonix.com/43779.html
Availability details for
Quantity
  • Availability
  • Add to Cart