SY3H3D67FC38 Synthonix Corporation

Methyl tetrahydro-2H-pyran-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 84355-44-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/43819.html

Safety Information

MSDS https://synthonix.com/43819.html
Availability details for
Quantity
  • Availability
  • Add to Cart