SY3H3D67FC40 Synthonix Corporation

(S)-Methyl 1-methylpyrrolidine-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 27957-91-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/43852.html

Safety Information

MSDS https://synthonix.com/43852.html
Availability details for
Quantity
  • Availability
  • Add to Cart