SY3H3D67FC44 Synthonix Corporation

Methyl piperidine-1-carbodithioate

  • MDL number No MDL found
  • Cas Number : 698-17-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/43860.html

Safety Information

MSDS https://synthonix.com/43860.html
Availability details for
Quantity
  • Availability
  • Add to Cart