SY3H3D67FC47 Synthonix Corporation

Methyl 1-methylpiperazine-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 405513-08-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43879.html

Safety Information

MSDS https://synthonix.com/43879.html
Availability details for
Quantity
  • Availability
  • Add to Cart