SY3H3D67FC4B Synthonix Corporation

trans-4-Methoxycyclohexanol

  • MDL number No MDL found
  • Cas Number : 22188-03-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43889.html

Safety Information

MSDS https://synthonix.com/43889.html
Availability details for
Quantity
  • Availability
  • Add to Cart