SY3H3D67FC4F Synthonix Corporation

1-Methyl-2-piperidinemethanol

  • MDL number No MDL found
  • Cas Number : 20845-34-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/43912.html

Safety Information

MSDS https://synthonix.com/43912.html
Availability details for
Quantity
  • Availability
  • Add to Cart