SY3H3D67FC53 Synthonix Corporation

(S)-(1-Methylpiperidin-2-yl)methanol

  • MDL number No MDL found
  • Cas Number : 136030-04-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43917.html

Safety Information

MSDS https://synthonix.com/43917.html
Availability details for
Quantity
  • Availability
  • Add to Cart