SY3H3D67FC55 Synthonix Corporation

Methyl 2,2-diethoxyacetimidate

  • Cas Number : 76742-48-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43935.html

Safety Information

MSDS https://synthonix.com/43935.html
Availability details for
Quantity
  • Availability
  • Add to Cart