SY3H3D67FC5B Synthonix Corporation

Methyl 2,6-dichloro-5-fluoronicotinate

  • MDL number No MDL found
  • Cas Number : 189281-66-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/44452.html

Safety Information

MSDS https://synthonix.com/44452.html
Availability details for
Quantity
  • Availability
  • Add to Cart