SY3H3D67FC5C Synthonix Corporation

4-Methoxy-5-nitro-2-(trifluoromethyl)pyridine

  • MDL number No MDL found
  • Cas Number : 1588441-18-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/44818.html

Safety Information

MSDS https://synthonix.com/44818.html
Availability details for
Quantity
  • Availability
  • Add to Cart