SY3H3D67FC60 Synthonix Corporation

Methyl 4,6-dibromopicolinate

  • MDL number No MDL found
  • Cas Number : 1206248-47-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/44931.html

Safety Information

MSDS https://synthonix.com/44931.html
Availability details for
Quantity
  • Availability
  • Add to Cart