SY3H3D67FC65 Synthonix Corporation

Methyl 4,5-dichloropicolinate

  • Cas Number : 1256834-28-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/44958.html

Safety Information

MSDS https://synthonix.com/44958.html
Availability details for
Quantity
  • Availability
  • Add to Cart