SY3H3D67FC76 Synthonix Corporation

3-(5-Methylisoxazol-3-yl)-3-oxopropanenitrile

  • Cas Number : 130371-64-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/45231.html

Safety Information

MSDS https://synthonix.com/45231.html
Availability details for
Quantity
  • Availability
  • Add to Cart