SY3H3D67FC77 Synthonix Corporation

4-Methoxy-2-oxo-1,2-dihydropyridine-3-carbonitrile

  • MDL number MFCD00975442
  • Cas Number : 21642-98-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Very pale yellow - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45232.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45232.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart