SY3H3D67FC7A Synthonix Corporation

Methyl 2-amino-4,6-dichloronicotinate

  • MDL number No MDL found
  • Cas Number : 1044872-40-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/45235.html

Safety Information

MSDS https://synthonix.com/45235.html
Availability details for
Quantity
  • Availability
  • Add to Cart