SY3H3D67FC7B Synthonix Corporation

Methyl 4-amino-3,6-dichloropicolinate

  • MDL number No MDL found
  • Cas Number : 350601-39-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/45236.html

Safety Information

MSDS https://synthonix.com/45236.html
Availability details for
Quantity
  • Availability
  • Add to Cart