SY3H3D67FC80 Synthonix Corporation

5-Methylpyrimidine-4,6-dicarboxylic acid

  • MDL number No MDL found
  • Cas Number : 2010958-16-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/45257.html

Safety Information

MSDS https://synthonix.com/45257.html
Availability details for
Quantity
  • Availability
  • Add to Cart