SY3H3D67FC81 Synthonix Corporation

2-Methoxy-5-nitronicotinic acid

  • MDL number No MDL found
  • Cas Number : 1020635-54-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/45259.html

Safety Information

MSDS https://synthonix.com/45259.html
Availability details for
Quantity
  • Availability
  • Add to Cart