SY3H3D67FC84 Synthonix Corporation

6-Methyl-3H-thieno[2,3-d]pyrimidin-4-one

  • MDL number No MDL found
  • Cas Number : 108831-66-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/45266.html

Safety Information

MSDS https://synthonix.com/45266.html
Availability details for
Quantity
  • Availability
  • Add to Cart