SY3H3D67FC86 Synthonix Corporation

6-Methylthiazolo[4,5-b]pyridine-2-thiol

  • MDL number No MDL found
  • Cas Number : 586409-05-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/45270.html

Safety Information

MSDS https://synthonix.com/45270.html
Availability details for
Quantity
  • Availability
  • Add to Cart