SY3H3D67FC89 Synthonix Corporation

Methyl 5-chloropyridine-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 132308-19-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/45366.html

Safety Information

MSDS https://synthonix.com/45366.html
Availability details for
Quantity
  • Availability
  • Add to Cart