SY3H3D67FC8A Synthonix Corporation

Methyl 4-chloronicotinate hydrochloride

  • MDL number No MDL found
  • Cas Number : 1351479-18-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/45369.html

Safety Information

MSDS https://synthonix.com/45369.html
Availability details for
Quantity
  • Availability
  • Add to Cart