SY3H3D67FC8E Synthonix Corporation

Methyl 5-bromo-4-hydroxynicotinate

  • MDL number No MDL found
  • Cas Number : 1175512-08-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/45424.html

Safety Information

MSDS https://synthonix.com/45424.html
Availability details for
Quantity
  • Availability
  • Add to Cart