SY3H3D67FC8F Synthonix Corporation

Methyl 3-chloro-6-oxo-1,6-dihydropyridine-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 1214361-05-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/45440.html

Safety Information

MSDS https://synthonix.com/45440.html
Availability details for
Quantity
  • Availability
  • Add to Cart