SY3H3D67FC92 Synthonix Corporation

6-Methyl-4-(trifluoromethyl)pyridin-2(1H)-one

  • MDL number No MDL found
  • Cas Number : 22123-19-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/45506.html

Safety Information

MSDS https://synthonix.com/45506.html
Availability details for
Quantity
  • Availability
  • Add to Cart