SY3H3D67FC96 Synthonix Corporation

Methyl 6-amino-5-bromonicotinate

  • Cas Number : 180340-70-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/45638.html

Safety Information

MSDS https://synthonix.com/45638.html
Availability details for
Quantity
  • Availability
  • Add to Cart