SY3H3D67FC98 Synthonix Corporation

Methyl 5-(bromomethyl)pyrazine-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 193966-70-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/45643.html

Safety Information

MSDS https://synthonix.com/45643.html
Availability details for
Quantity
  • Availability
  • Add to Cart