SY3H3D67FC99 Synthonix Corporation

Methyl 6-bromo-5-methylpyrazine-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 1166827-91-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/45644.html

Safety Information

MSDS https://synthonix.com/45644.html
Availability details for
Quantity
  • Availability
  • Add to Cart