SY3H3D67FC9B Synthonix Corporation

Methyl 5-amino-6-chloronicotinate

  • Cas Number : 211915-96-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/45653.html

Safety Information

MSDS https://synthonix.com/45653.html
Availability details for
Quantity
  • Availability
  • Add to Cart