SY3H3D67FC9D Synthonix Corporation

Methyl 6-amino-4-chloronicotinate

  • Cas Number : 1260666-60-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/45655.html

Safety Information

MSDS https://synthonix.com/45655.html
Availability details for
Quantity
  • Availability
  • Add to Cart