SY3H3D67FCA2 Synthonix Corporation

2-Methoxy-6-(trifluoromethyl)pyridin-3-amine

  • Cas Number : 944900-09-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/45712.html

Safety Information

MSDS https://synthonix.com/45712.html
Availability details for
Quantity
  • Availability
  • Add to Cart