SY3H3D67FCA3 Synthonix Corporation

3-Methoxy-4-(trifluoromethyl)pyridin-2-amine

  • MDL number No MDL found
  • Cas Number : 1227581-88-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/45713.html

Safety Information

MSDS https://synthonix.com/45713.html
Availability details for
Quantity
  • Availability
  • Add to Cart