SY3H3D67FCA4 Synthonix Corporation

3-Methoxy-5-(trifluoromethyl)pyridin-2-amine

  • Cas Number : 1227603-09-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/45714.html

Safety Information

MSDS https://synthonix.com/45714.html
Availability details for
Quantity
  • Availability
  • Add to Cart