SY3H3D67FCA5 Synthonix Corporation

3-Methoxy-6-(trifluoromethyl)pyridin-2-amine

  • MDL number No MDL found
  • Cas Number : 1228898-19-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/45715.html

Safety Information

MSDS https://synthonix.com/45715.html
Availability details for
Quantity
  • Availability
  • Add to Cart