SY3H3D67FCA6 Synthonix Corporation

4-Methoxy-5-(trifluoromethyl)pyridin-2-amine

  • MDL number No MDL found
  • Cas Number : 1227571-99-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/45716.html

Safety Information

MSDS https://synthonix.com/45716.html
Availability details for
Quantity
  • Availability
  • Add to Cart