SY3H3D67FCA7 Synthonix Corporation

6-Methoxy-4-(trifluoromethyl)pyridin-2-amine

  • MDL number No MDL found
  • Cas Number : 1598267-75-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/45717.html

Safety Information

MSDS https://synthonix.com/45717.html
Availability details for
Quantity
  • Availability
  • Add to Cart