SY3H3D67FCAD Synthonix Corporation

2-Methyl-7H-pyrrolo[2,3-d]pyrimidine

  • MDL number No MDL found
  • Cas Number : 89792-07-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/45731.html

Safety Information

MSDS https://synthonix.com/45731.html
Availability details for
Quantity
  • Availability
  • Add to Cart