SY3H3D67FCB0 Synthonix Corporation

2-Methyl-1H-pyrrolo[2,3-d]pyrimidin-4(7H)-one

  • MDL number No MDL found
  • Cas Number : 89792-11-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/45742.html

Safety Information

MSDS https://synthonix.com/45742.html
Availability details for
Quantity
  • Availability
  • Add to Cart