SY3H3D67FCB4 Synthonix Corporation

2-Methyl-4-(methylthio)-6-oxo-1,6-dihydropyrimidine-5-carbonitrile

  • MDL number No MDL found
  • Cas Number : 15908-63-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/45754.html

Safety Information

MSDS https://synthonix.com/45754.html
Availability details for
Quantity
  • Availability
  • Add to Cart