SY3H3D67FCB8 Synthonix Corporation

(6-Methyl-5-(trifluoromethyl)pyridin-3-yl)boronic acid

  • MDL number No MDL found
  • Cas Number : 1889221-15-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/45824.html

Safety Information

MSDS https://synthonix.com/45824.html
Availability details for
Quantity
  • Availability
  • Add to Cart