SY3H3D67FCC4 Synthonix Corporation

Methyl 4-oxo-1,4-dihydropyridine-2-carboxylate

  • MDL number MFCD13174828
  • Cas Number : 98321-25-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45849.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45849.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart